Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitroaniline-4-sulfonic Acid 94.0+%, TCI America™

Click to view available options
Quantity:
25g
712-24-3
C6H6N2O5S
218.183
MFCD00059185
ZNMGCJDUZADKPW-UHFFFAOYSA-N
4-Amino-2-nitrobenzenesulfonic Acid
69735
C1=CC(=C(C=C1N)[N+](=O)[O-])S(=O)(=O)O
712-24-3
C6H6N2O5S
218.183
MFCD00059185
ZNMGCJDUZADKPW-UHFFFAOYSA-N
4-Amino-2-nitrobenzenesulfonic Acid
69735
C1=CC(=C(C=C1N)[N+](=O)[O-])S(=O)(=O)O
Quantity 25g
Color Undesignated
Formula Weight 218.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥94.0% (HPLC)
Chemical Name or Material 3-Nitroaniline-4-sulfonic Acid

EINECSNumber : (3)-2078

TSCA : Yes