Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Nitroaniline, 98%

Manufacturer:  Alfa Aesar™ A1062922

Quantity 100g

 View more versions of this product

Catalog No. AAA1062922


  Check Availability
Add to cart

Specifications

Description

Applications
3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.

Solubility
It is soluble in water 0.1 g/100 ml (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids, acid chlorides, acid anhydrides, chloroformates, strong oxidizing agents.
Description & Specifications

Chemical Identifiers

99-09-2
138.13
UN1661
14,6582
C6H6N2O2
MFCD00007782
636962
3-Nitroaniline

Specifications

306°C
100g
C6H6N2O2
MFCD00007782
636962
3-Nitroaniline
112° to 114°C
99-09-2
138.13
UN1661
14,6582
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H310-H330-H373-H302
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.

GHS P Statement
P260-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROANILINES

RTECSNumber : BY6825000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit