Learn More
3-Nitroaniline, 98%, Thermo Scientific Chemicals
Description
3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.
Solubility
It is soluble in water 0.1 g/100 ml (20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids, acid chlorides, acid anhydrides, chloroformates, strong oxidizing agents.
Specifications
Melting Point | 112°C to 114°C |
Density | 0.901 |
Boiling Point | 306°C |
Quantity | 500 g |
UN Number | UN1661 |
Beilstein | 636962 |
Merck Index | 14,6582 |
Solubility Information | It is soluble in water 0.1g/100 ml (20°C). |
Formula Weight | 138.13 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H301+H311+H331-H373 |
Hazard Statement | GHS H Statement H310-H330-H373-H302 Fatal in contact with skin. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. |
Precautionary Statement | P260-P262-P264b-P270-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROANILINES |
EINECSNumber | 202-729-1 |
RTECSNumber | BY6825000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.