Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog Number p-172166
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
99-61-6
C7H5NO3
151.12
MFCD00007249
ZETIVVHRRQLWFW-UHFFFAOYSA-N
m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde
7449
3-nitrobenzaldehyde
C1=CC(=CC(=C1)[N+](=O)[O-])C=O
99-61-6
C7H5NO3
151.12
MFCD00007249
ZETIVVHRRQLWFW-UHFFFAOYSA-N
m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde
7449
3-nitrobenzaldehyde
C1=CC(=CC(=C1)[N+](=O)[O-])C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 56.0°C to 59.0°C
Color Brown-Yellow to Yellow
Boiling Point 285.0°C to 290.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula O2NC6H4CHO
Quantity 100 g
Beilstein 07, 250
Merck Index 14, 6587
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, chloroformm and ether
Formula Weight 151.12
Percent Purity 99%
Physical Form Granular Powder
Chemical Name or Material 3-Nitrobenzaldehyde, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear
EINECSNumber 202-772-6
RTECSNumber CU7250000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit