Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrobenzamide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
645-09-0
C7H6N2O3
166.14
MFCD00007984
KWAYEPXDGHYGRW-UHFFFAOYSA-N
m-nitrobenzamide,benzamide, 3-nitro,benzamide, m-nitro,3-nitro-benzamide,3-nitrophenylcarboxamide,acmc-209nmr,3-nitrobenzamide,dsstox_cid_5732,dsstox_rid_77897,dsstox_gsid_25732
12576
3-nitrobenzamide
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)N
645-09-0
C7H6N2O3
166.14
MFCD00007984
KWAYEPXDGHYGRW-UHFFFAOYSA-N
m-nitrobenzamide,benzamide, 3-nitro,benzamide, m-nitro,3-nitro-benzamide,3-nitrophenylcarboxamide,acmc-209nmr,3-nitrobenzamide,dsstox_cid_5732,dsstox_rid_77897,dsstox_gsid_25732
12576
3-nitrobenzamide
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)N
Melting Point 140°C to 144°C
Infrared Spectrum Authentic
Quantity 25g
Color Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula O2NC6H4CONH2
Beilstein 09, 381
Solubility Information Solubility in water: slightly soluble
Formula Weight 166.14
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzamide, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

EINECSNumber : 211-431-0

RTECSNumber : CV5601250

TSCA : TSCA