Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Nitrobenzeneboronic acid L-tartaric acid ester, 1M soln. in THF

Manufacturer:  Alfa Aesar™ H55216AA

Quantity 5mL

 View more versions of this product

Catalog No. AAH55216AA


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

467443-01-2
MFCD09750458
3-Nitrophenylboronic acid L-tartaric acid ester, 1M soln. in THF
280.99
UN2056
3-Nitrobenzeneboronic acid L-tartaric acid ester

Specifications

1M soln. in THF
Moisture Sensitive
5mL
Liquid
280.99
UN2056
3-Nitrobenzeneboronic acid L-tartaric acid ester
1.053g/mL
Colorless to brown
467443-01-2
1.4508
MFCD09750458
3-Nitrophenylboronic acid L-tartaric acid ester, 1M soln. in THF

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: TETRAHYDROFURAN

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit