Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrobenzenesulfonamide, 99%, Thermo Scientific™

Catalog Number AAH5583103
Click to view available options
Quantity:
1 g
5 g
25 g
121-52-8
C6H6N2O4S
202.184
MFCD00007935
TXTQURPQLVHJRE-UHFFFAOYSA-N
benzenesulfonamide, 3-nitro, m-nitrobenzenesulfonamide, 3-nitrobenzolesulfamide, 3-nitrobenzenesulphonamide, 3-nitrobenzene-1-sulfonamide, m-nitrobenzenesulphonamide, 3-nitro-benzenesulfonamide, benzenesulfonamide, m-nitro, 3-nitrobenzene sulfonamide, m-nitrobenzolsulfonamid
67138
3-nitrobenzenesulfonamide
C1=CC(=CC(=C1)S(=O)(=O)N)[N+](=O)[O-]
121-52-8
C6H6N2O4S
202.184
MFCD00007935
TXTQURPQLVHJRE-UHFFFAOYSA-N
benzenesulfonamide, 3-nitro, m-nitrobenzenesulfonamide, 3-nitrobenzolesulfamide, 3-nitrobenzenesulphonamide, 3-nitrobenzene-1-sulfonamide, m-nitrobenzenesulphonamide, 3-nitro-benzenesulfonamide, benzenesulfonamide, m-nitro, 3-nitrobenzene sulfonamide, m-nitrobenzolsulfonamid
67138
3-nitrobenzenesulfonamide
C1=CC(=CC(=C1)S(=O)(=O)N)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 166°C to 168°C
Color White
Quantity 1 g
Formula Weight 202.19
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Nitrobenzenesulfonamide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 204-477-8
TSCA Yes
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit