Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrobenzoic Acid, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
5kg
121-92-6
C7H5NO4
167.12
MFCD00007251
AFPHTEQTJZKQAQ-UHFFFAOYSA-N
3-nitrobenzoic acid,m-nitrobenzoic acid,benzoic acid, 3-nitro,benzoic acid, m-nitro,meta-nitrobenzoic acid,metanitrobenzoic acid,m-nitrobenzenecarboxylic acid,3-nitro-benzoic acid,ccris 2335,nitrobenzoic 3-acid
8497
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O
121-92-6
C7H5NO4
167.12
MFCD00007251
AFPHTEQTJZKQAQ-UHFFFAOYSA-N
3-nitrobenzoic acid,m-nitrobenzoic acid,benzoic acid, 3-nitro,benzoic acid, m-nitro,meta-nitrobenzoic acid,metanitrobenzoic acid,m-nitrobenzenecarboxylic acid,3-nitro-benzoic acid,ccris 2335,nitrobenzoic 3-acid
8497
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O
Melting Point 139.0°C to 143.0°C
Flash Point 190°C
Color Green to Yellow
Quantity 100g
Packaging Plastic bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula O2NC6H4CO2H
Beilstein 09,376
Fieser 13,231
Merck Index 14,6589
Solubility Information Solubility in water: 3g/L (25°C). Other solubilities: 1g/3mL alcohol - 1g/2mL methanol, 1g/4mL ether - 1g/2.5mL acetone, 1g/18mL chloroform, slightly soluble in benzene and carbon disulfide
Formula Weight 167.12
Physical Form Crystals or Powder
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzoic acid
Show More Show Less

GHS H Statement:
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.