Alfa Aesar™ 3-Nitrobenzyl alcohol, 99+%

Manufacturer: Alfa Aesar™ 4444503

Quantity 1g

 View more versions of this product

Catalog No. AA4444503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 619-25-0
    Molecular Formula C7H7NO3
    Formula Weight 153.14
    MDL Number MFCD00007273
    Beilstein 2044769
    Chemical Name or Material 3-Nitrobenzyl alcohol

    Specifications

    Formula Weight 153.14
    Physical Form Solid
    Molecular Formula C7H7NO3
    RTECS Number DP0657000
    Flash Point >110°C (230°F)
    Boiling Point 175° to 180°C (3mmHg)
    Color Yellow
    Quantity 1g
    Refractive Index 1.5730
    TSCA TSCA
    MDL Number MFCD00007273
    CAS 619-25-0
    Beilstein 2044769
    Density 1.300g/mL
    Melting Point 29° to 33°C
    Chemical Name or Material 3-Nitrobenzyl alcohol
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit