Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrobenzylamine hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
26177-43-5
C7H8N2O2·HCl
188.62
MFCD00012858
DLZXLCHQWOZGSE-UHFFFAOYSA-N
3-nitrobenzylamine hydrochloride,3-nitrophenyl methanamine hydrochloride,3-nitrobenzylamine hcl,3-nitrobenzylammonium hydrochloride,benzenemethanamine, 3-nitro-, monohydrochloride,1-3-nitrophenyl methanamine hydrochloride,pubchem7485,chembl13978,m-nitrobenzylamine hydrochloride,3-nitrobenzyl amine hydrochloride
2724030
(3-nitrophenyl)methanamine;hydrochloride
C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl
26177-43-5
C7H8N2O2·HCl
188.62
MFCD00012858
DLZXLCHQWOZGSE-UHFFFAOYSA-N
3-nitrobenzylamine hydrochloride,3-nitrophenyl methanamine hydrochloride,3-nitrobenzylamine hcl,3-nitrobenzylammonium hydrochloride,benzenemethanamine, 3-nitro-, monohydrochloride,1-3-nitrophenyl methanamine hydrochloride,pubchem7485,chembl13978,m-nitrobenzylamine hydrochloride,3-nitrobenzyl amine hydrochloride
2724030
(3-nitrophenyl)methanamine;hydrochloride
C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl
Packaging Glass bottle
Melting Point 226°C to 231°C
Infrared Spectrum Authentic
Quantity 1g
Color Tan to White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula O2NC6H4CH2NH2·HCl
Beilstein 12, 1076
Formula Weight 188.62
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3-Nitrobenzylamine hydrochloride, 97%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 247-502-8

TSCA : TSCA