Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrobenzylamine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number AC184730050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
26177-43-5
C7H8N2O2·HCl
188.62
MFCD00012858
DLZXLCHQWOZGSE-UHFFFAOYSA-N
3-nitrobenzylamine hydrochloride, 3-nitrophenyl methanamine hydrochloride, 3-nitrobenzylamine hcl, 3-nitrobenzylammonium hydrochloride, benzenemethanamine, 3-nitro-, monohydrochloride, 1-3-nitrophenyl methanamine hydrochloride, pubchem7485, chembl13978, m-nitrobenzylamine hydrochloride, 3-nitrobenzyl amine hydrochloride
2724030
(3-nitrophenyl)methanamine;hydrochloride
C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl
26177-43-5
C7H8N2O2·HCl
188.62
MFCD00012858
DLZXLCHQWOZGSE-UHFFFAOYSA-N
3-nitrobenzylamine hydrochloride, 3-nitrophenyl methanamine hydrochloride, 3-nitrobenzylamine hcl, 3-nitrobenzylammonium hydrochloride, benzenemethanamine, 3-nitro-, monohydrochloride, 1-3-nitrophenyl methanamine hydrochloride, pubchem7485, chembl13978, m-nitrobenzylamine hydrochloride, 3-nitrobenzyl amine hydrochloride
2724030
(3-nitrophenyl)methanamine;hydrochloride
C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 226°C to 231°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula O2NC6H4CH2NH2·HCl
Quantity 5 g
Beilstein 12, 1076
Formula Weight 188.62
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Nitrobenzylamine hydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 247-502-8
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit