Alfa Aesar™ 3-Nitrocinnamic acid, predominantly trans, 98%

Manufacturer: Alfa Aesar™ A1219922

Quantity 100g

 View more versions of this product

Catalog No. AAA1219922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 555-68-0
    Molecular Formula C9H7NO4
    Formula Weight 193.16
    MDL Number MFCD00007277
    Beilstein 1872713
    Merck Index 14,6595
    Chemical Name or Material 3-Nitrocinnamic acid, predominantly trans

    Specifications

    Formula Weight 193.16
    TSCA TSCA
    Merck Index 14,6595
    Health Hazard 1 Warning
    Melting Point 202° to 206°C
    Quantity 100g
    Molecular Formula C9H7NO4
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    RTECS Number GE0352000
    Odor Odorless
    MDL Number MFCD00007277
    CAS 555-68-0
    Beilstein 1872713
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 3-Nitrocinnamic acid, predominantly trans
    Assay 98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit