Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Nitrophenol, 98+%

Click to view available options
Quantity:
10g
50g
250g
554-84-7
C6H5NO3
139.11
MFCD00007240
RTZZCYNQPHTPPL-UHFFFAOYSA-N
m-nitrophenol,3-hydroxynitrobenzene,m-hydroxynitrobenzene,phenol, 3-nitro,phenol, m-nitro,m-nitrofenol,meta-nitrophenol,3-nitro-phenol,m-nitrofenol czech,1-hydroxy-3-nitrobenzene
11137
CHEBI:34346
3-nitrophenol
C1=CC(=CC(=C1)O)[N+](=O)[O-]
554-84-7
C6H5NO3
139.11
MFCD00007240
RTZZCYNQPHTPPL-UHFFFAOYSA-N
m-nitrophenol,3-hydroxynitrobenzene,m-hydroxynitrobenzene,phenol, 3-nitro,phenol, m-nitro,m-nitrofenol,meta-nitrophenol,3-nitro-phenol,m-nitrofenol czech,1-hydroxy-3-nitrobenzene
11137
CHEBI:34346
3-nitrophenol
C1=CC(=CC(=C1)O)[N+](=O)[O-]
Applications
3-Nitrophenol is used as indicator in 0.3% Soln in 50% alcohol ph: 6.8 Colorless, 8.6 Yellow. It is used in the synthesis of RAGE inhibitors via the etherification of phenols. Also used in the synthesis of kinase inhibitors against epidermal growth factor receptor T790M mutation.

Solubility
Soluble in water (30 mg/ml), ether, alcohol, acetone, and ethanol (700 mg/ml).

Notes
Store in a tightly closed container. Keep from contact with oxidizing materials. Store in a cool, dry, well-ventilated area away from incompatible substances.
Density 1.49
Boiling Point 194°C (70mmHg)
Melting Point 93°C to 98°C
Quantity 50g
Flash Point >100°C (212°F)
UN Number UN1663
Beilstein 1907946
Merck Index 14,6619
Solubility Information Soluble in water (30mg/ml),ether,alcohol,acetone,and ethanol (700mg/ml).
Formula Weight 139.11
Percent Purity ≥98%
Chemical Name or Material 3-Nitrophenol
Show More Show Less

GHS H Statement
H318-H373-H302-H312-H335-H315
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.

P260-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P314-P330-P332+P313-P362-P501c

H302-H315-H318-H373-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROPHENOLS

EINECSNumber : 209-073-5

RTECSNumber : SM1925000

TSCA : Yes

Recommended Storage : Ambient temperatures