Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrophenol, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
554-84-7
C6H5NO3
139.11
RTZZCYNQPHTPPL-UHFFFAOYSA-N
m-nitrophenol,3-hydroxynitrobenzene,m-hydroxynitrobenzene,phenol, 3-nitro,phenol, m-nitro,m-nitrofenol,meta-nitrophenol,3-nitro-phenol,m-nitrofenol czech,1-hydroxy-3-nitrobenzene
11137
CHEBI:34346
3-nitrophenol
C1=CC(=CC(=C1)O)[N+](=O)[O-]
554-84-7
C6H5NO3
139.11
RTZZCYNQPHTPPL-UHFFFAOYSA-N
m-nitrophenol,3-hydroxynitrobenzene,m-hydroxynitrobenzene,phenol, 3-nitro,phenol, m-nitro,m-nitrofenol,meta-nitrophenol,3-nitro-phenol,m-nitrofenol czech,1-hydroxy-3-nitrobenzene
11137
CHEBI:34346
3-nitrophenol
C1=CC(=CC(=C1)O)[N+](=O)[O-]
Infrared Spectrum Authentic
Melting Point 94.0°C to 98.0°C
Color Yellow to Brown
Density 1.4900g/mL
Boiling Point 194.0°C (70.0 mmHg)
Quantity 50g
Packaging Glass bottle
Specific Gravity 1.49
Assay Percent Range 98.5% min. (GC)
Linear Formula O2NC6H4OH
Beilstein 06,222
Merck Index 15,6706
Solubility Information Solubility in water: 13.5g/L (25°C). Other solubilities: soluble 700g/L in ethanol (20°C),soluble in benzene,diethyl ether,alcohol,,hot and diluted solutions,insoluble in petroleum ehter
Formula Weight 139.11
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 3-Nitrophenol
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear prote

GHS Signal Word: Danger

EINECSNumber : 209-073-5