Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrophenylhydrazine Hydrochloride 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
636-95-3
C6H8ClN3O2
189.599
MFCD00012939
BKOYKMLGFFASBG-UHFFFAOYSA-N
3-nitrophenylhydrazine hydrochloride,3-nitrophenyl hydrazine hydrochloride,3-nitrophenylhydrazinium 1+ chloride,hydrazine, 3-nitrophenyl-, monohydrochloride,m-nitrophenylhydrazine hydrochloride,pubchem20000,acmc-1b1e3,ksc491e2l,m-nitrophenylhydrazine hcl
5464285
(3-nitrophenyl)hydrazine;hydrochloride
C1=CC(=CC(=C1)[N+](=O)[O-])NN.Cl
636-95-3
C6H8ClN3O2
189.599
MFCD00012939
BKOYKMLGFFASBG-UHFFFAOYSA-N
3-nitrophenylhydrazine hydrochloride,3-nitrophenyl hydrazine hydrochloride,3-nitrophenylhydrazinium 1+ chloride,hydrazine, 3-nitrophenyl-, monohydrochloride,m-nitrophenylhydrazine hydrochloride,pubchem20000,acmc-1b1e3,ksc491e2l,m-nitrophenylhydrazine hcl
5464285
(3-nitrophenyl)hydrazine;hydrochloride
C1=CC(=CC(=C1)[N+](=O)[O-])NN.Cl
Sulfated Ash 0.05% max.
Infrared Spectrum Authentic
Melting Point 206°C to 210°C
Quantity 5g
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula O2NC6H4NHNH2·HCl
Beilstein 15, 460
Formula Weight 189.6
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Nitrophenylhydrazine hydrochloride, 98%
Show More Show Less

GHS H Statement:
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Risk of explosion if heated under confinement.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-270-6

TSCA : TSCA