Alfa Aesar™ 3-Nitrophthalonitrile, 99%

Manufacturer: Alfa Aesar™ B2526914

Quantity 25g

 View more versions of this product

Catalog No. AAB2526914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 51762-67-5
    Molecular Formula C8H3N3O2
    Formula Weight 173.13
    MDL Number MFCD00191558
    Beilstein 2263686
    Synonym 2,3-Dicyanonitrobenzene
    Chemical Name or Material 3-Nitrophthalonitrile

    Specifications

    Formula Weight 173.13
    Molecular Formula C8H3N3O2
    MDL Number MFCD00191558
    Melting Point 162° to 165°C
    CAS 51762-67-5
    Quantity 25g
    Synonym 2,3-Dicyanonitrobenzene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number CZ1953200
    Beilstein 2263686
    Chemical Name or Material 3-Nitrophthalonitrile
    Assay 99%

    Description

    Applications
    3-Nitrophthalonitrile is a useful dye for biological research purposes. It is used as an intermediate in the manufacture of pharmaceuticals, chemicals, dyestuff and phthalocyanine blue pigments.

    Solubility
    Soluble in methanol. Insoluble in water.

    Notes
    Incombatible with strong oxidizing agents and strong bases.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit