Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitropropionic acid, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
504-88-1
C3H5NO4
119.076
WBLZUCOIBUDNBV-UHFFFAOYSA-N
3-nitropropionic acid,3-nitropropanoic acid,bovinocidin,hiptagenic acid,propanoic acid, 3-nitro,beta-nitropropionic acid,3-nitropropionate,propionic acid, 3-nitro,beta-nitropropanoic acid,3-np acid
1678
CHEBI:16348
C(C[N+](=O)[O-])C(=O)O
This item is not returnable. View return policy
504-88-1
C3H5NO4
119.076
WBLZUCOIBUDNBV-UHFFFAOYSA-N
3-nitropropionic acid,3-nitropropanoic acid,bovinocidin,hiptagenic acid,propanoic acid, 3-nitro,beta-nitropropionic acid,3-nitropropionate,propionic acid, 3-nitro,beta-nitropropanoic acid,3-np acid
1678
CHEBI:16348
C(C[N+](=O)[O-])C(=O)O

Irreversible mitochondrial respiratory complex II inhibitor

Irreversible mitochondrial respiratory complex II (succinate dehydrogenase) inhibitor; induces autophagy in SH-SY5Y cells. Recapitulates Huntington's disease-like pathology and symptoms in primate and rodent models.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 119.08
Percent Purity >97%
Chemical Name or Material 3-Nitropropionic acid

Recommended Storage : Store at +4°C