Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrostyrene, 97%, ACROS Organics™

Click to view available options
Quantity:
2.5g
10g
Packaging:
Glass bottle
586-39-0
C8H7NO2
149.15
MFCD00007276
SYZVQXIUVGKCBJ-UHFFFAOYSA-N
3-nitrostyrene,1-nitro-3-vinylbenzene,m-nitrostyrene,benzene, 1-ethenyl-3-nitro,styrene, m-nitro,1-nitro-3-vinyl-benzene,3-nitrophenyl ethene,b-nitrostyrene,m-vinylnitrobenzene,3-ethenylnitrobenzene
68514
1-ethenyl-3-nitrobenzene
C=CC1=CC(=CC=C1)[N+](=O)[O-]
586-39-0
C8H7NO2
149.15
MFCD00007276
SYZVQXIUVGKCBJ-UHFFFAOYSA-N
3-nitrostyrene,1-nitro-3-vinylbenzene,m-nitrostyrene,benzene, 1-ethenyl-3-nitro,styrene, m-nitro,1-nitro-3-vinyl-benzene,3-nitrophenyl ethene,b-nitrostyrene,m-vinylnitrobenzene,3-ethenylnitrobenzene
68514
1-ethenyl-3-nitrobenzene
C=CC1=CC(=CC=C1)[N+](=O)[O-]
Melting Point -5°C
Flash Point 107°C
Specific Gravity 1.07
Refractive Index 1.5820 to 1.584
Stabilizer approx. 0.1% 3, 5-di-tert-butylcatechol
Quantity 2.5g
Infrared Spectrum Authentic
Density 1.0700g/mL
Packaging Glass bottle
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula H2C=CHC6H4NO2
Beilstein 05, 478
Formula Weight 149.15
Physical Form Powder or Needles
Percent Purity ≥96.0% (GC)
Chemical Name or Material 3-Nitrostyrene
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning