Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-O-Benzoyl-4,6-O-isopropylidene-D-glucal, 97%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
58871-20-8
C16H18O5
290.32
MFCD22988992
RADJUQJHMUFUJI-UHFFFAOYNA-N
3-o-benzoyl-4,6-o-isopropylidene-d-glucal, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl benzoate
92135621
[(4aR,8R,8aS)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl] benzoate
CC1(C)OCC2OC=CC(OC(=O)C3=CC=CC=C3)C2O1
58871-20-8
C16H18O5
290.32
MFCD22988992
RADJUQJHMUFUJI-UHFFFAOYNA-N
3-o-benzoyl-4,6-o-isopropylidene-d-glucal, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl benzoate
92135621
[(4aR,8R,8aS)-2,2-dimethyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-yl] benzoate
CC1(C)OCC2OC=CC(OC(=O)C3=CC=CC=C3)C2O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Quantity 250mg
Formula Weight 290.31
Percent Purity 97%
Chemical Name or Material 3-O-Benzoyl-4,6-O-isopropylidene-D-glucal

TSCA : No

Recommended Storage : Keep cold