Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Octanone, 98%, Thermo Scientific™

Click to view available options
Quantity:
100g
500g
106-68-3
C8H16O
128.215
MFCD00009515
RHLVCLIPMVJYKS-UHFFFAOYSA-N
3-octanone, ethyl amyl ketone, ethyl pentyl ketone, amyl ethyl ketone, n-octanone-3, ethyl n-amyl ketone, 3-oxooctane, ethyl n-pentyl ketone, ethyl n-amylketone, 3-octanone natural
246728
CHEBI:80946
octan-3-one
CCCCCC(=O)CC
106-68-3
C8H16O
128.215
MFCD00009515
RHLVCLIPMVJYKS-UHFFFAOYSA-N
3-octanone, ethyl amyl ketone, ethyl pentyl ketone, amyl ethyl ketone, n-octanone-3, ethyl n-amyl ketone, 3-oxooctane, ethyl n-pentyl ketone, ethyl n-amylketone, 3-octanone natural
246728
CHEBI:80946
octan-3-one
CCCCCC(=O)CC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Incompatible with strong oxidizing agents, strong reducing agents, strong bases.
Density 0.822
Melting Point -23°C
Boiling Point 167°C to 168°C
Flash Point 51°C (123°F)
UN Number UN1224
Beilstein 1700021
Merck Index 14,6084
Refractive Index 1.415
Quantity 500g
Solubility Information Insoluble in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 3-Octanone
Show More Show Less

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c

H226-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 203-423-0

RTECSNumber : RH1485000

TSCA : Yes

Recommended Storage : Ambient temperatures

RUO – Research Use Only