Alfa Aesar™ 3-Octanone, 98%

Manufacturer: Alfa Aesar™ A1211314

Quantity 25g

 View more versions of this product

Catalog No. AAA1211314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 106-68-3
    Molecular Formula C8H16O
    Formula Weight 128.22
    MDL Number MFCD00009515
    UN Number UN1224
    Beilstein 1700021
    Merck Index 14,6084
    Synonym n-Amyl ethyl ketone; Ethyl n-pentyl ketone
    Chemical Name or Material 3-Octanone

    Specifications

    Formula Weight 128.22
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 25g
    CAS 106-68-3
    Boiling Point 167° to 168°C
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number RH1485000
    MDL Number MFCD00009515
    Merck Index 14,6084
    Synonym n-Amyl ethyl ketone; Ethyl n-pentyl ketone
    Melting Point −23°C
    Flash Point 51°C (123°F)
    TSCA TSCA
    Molecular Formula C8H16O
    Beilstein 1700021
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
    UN Number UN1224
    Health Hazard 1 Warning
    Chemical Name or Material 3-Octanone
    Assay 98%

    Description

    Applications
    3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.

    Solubility
    Insoluble in water.

    Notes
    Store in cool, dry place in a tightly sealed container. Incompatible with strong oxidizing agents, strong reducing agents, strong bases.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit