Alfa Aesar™ 3-Octen-2-ol, 97%

Manufacturer: Alfa Aesar™ B2174106

Quantity 5g

 View more versions of this product

Catalog No. AAB2174106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76649-14-4
    Molecular Formula C8H16O
    Formula Weight 128.22
    MDL Number MFCD00036565
    Beilstein 1743648
    Chemical Name or Material 3-Octen-2-ol

    Specifications

    Formula Weight 128.22
    TSCA TSCA
    Molecular Formula C8H16O
    MDL Number MFCD00036565
    CAS 76649-14-4
    Beilstein 1743648
    Boiling Point 98°C (2mmHg)
    Quantity 5g
    Flash Point 71°C (159°F)
    Chemical Name or Material 3-Octen-2-ol
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit