Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Oxabicyclo[3.1.0]hexane-2,4-dione, ≥97%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
5617-74-3
C5H4O3
112.084
MFCD00126929
ZRMYHUFDVLRYPN-UHFFFAOYSA-N
3-oxabicyclo 3.1.0 hexane-2,4-dione,1,2-cyclopropanedicarboxylic anhydride,3-oxabicyclo 3.1.0 hexane-2,4-dione,oxabicyclohexanedione,pubchem18320,cyclopropane-1,2-dicarboxylic anhydride,2,4-dioxo-3-oxabicyclo 3.1.0 hexane,3-oxa-bicyclo 3.1.0 hexane-2,4-dione,3-oxabicyclo 3.1.0-hexane-2,4-dione,3-oxa-bicyclo 3.1.0-hexane-2,4-dione
2734721
3-oxabicyclo[3.1.0]hexane-2,4-dione
C1C2C1C(=O)OC2=O
5617-74-3
C5H4O3
112.084
MFCD00126929
ZRMYHUFDVLRYPN-UHFFFAOYSA-N
3-oxabicyclo 3.1.0 hexane-2,4-dione,1,2-cyclopropanedicarboxylic anhydride,3-oxabicyclo 3.1.0 hexane-2,4-dione,oxabicyclohexanedione,pubchem18320,cyclopropane-1,2-dicarboxylic anhydride,2,4-dioxo-3-oxabicyclo 3.1.0 hexane,3-oxa-bicyclo 3.1.0 hexane-2,4-dione,3-oxabicyclo 3.1.0-hexane-2,4-dione,3-oxa-bicyclo 3.1.0-hexane-2,4-dione
2734721
3-oxabicyclo[3.1.0]hexane-2,4-dione
C1C2C1C(=O)OC2=O
Quantity 1g
Density 1.50g/mL
Boiling Point 100°C to 102°C (5mmHg)
Color White
Melting Point 59°C to 61°C (literature)
Sensitivity Moisture Sensitive
Beilstein 113493
Formula Weight 112.08
Physical Form Powder
Percent Purity ≥97%
Chemical Name or Material 3-Oxabicyclo[3.1.0]hexane-2,4-dione
Show More Show Less