Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Oxabicyclo[3.1.0]hexane-2,4-dione, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
5617-74-3
C5H4O3
112.09
MFCD00126929
ZRMYHUFDVLRYPN-UHFFFAOYSA-N
3-oxabicyclo 3.1.0 hexane-2,4-dione,1,2-cyclopropanedicarboxylic anhydride,oxabicyclohexanedione,pubchem18320,cyclopropane-1,2-dicarboxylic anhydride,2,4-dioxo-3-oxabicyclo 3.1.0 hexane,3-oxa-bicyclo 3.1.0 hexane-2,4-dione,3-oxabicyclo 3.1.0-hexane-2,4-dione,3-oxa-bicyclo 3.1.0-hexane-2,4-dione
2734721
3-oxabicyclo[3.1.0]hexane-2,4-dione
C1C2C1C(=O)OC2=O
5617-74-3
C5H4O3
112.09
MFCD00126929
ZRMYHUFDVLRYPN-UHFFFAOYSA-N
3-oxabicyclo 3.1.0 hexane-2,4-dione,1,2-cyclopropanedicarboxylic anhydride,oxabicyclohexanedione,pubchem18320,cyclopropane-1,2-dicarboxylic anhydride,2,4-dioxo-3-oxabicyclo 3.1.0 hexane,3-oxa-bicyclo 3.1.0 hexane-2,4-dione,3-oxabicyclo 3.1.0-hexane-2,4-dione,3-oxa-bicyclo 3.1.0-hexane-2,4-dione
2734721
3-oxabicyclo[3.1.0]hexane-2,4-dione
C1C2C1C(=O)OC2=O
Boiling Point 100°C to 102°C
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
Quantity 5g
Melting Point 54°C to 60°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 112.09
Physical Form Powder, Crystals and/or Chunks
Percent Purity 98%
Chemical Name or Material 3-Oxabicyclo[3.1.0]hexane-2, 4-dione, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning