Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Oxocyclobutanecarboxylic acid, 98%

Click to view available options
Quantity:
5g
25g
23761-23-1
C5H6O3
114.1
MFCD00100900
IENOFRJPUPTEMI-UHFFFAOYSA-N
3-oxocyclobutanecarboxylic acid,3-oxo-cyclobutanecarboxylic acid,cyclobutanecarboxylic acid, 3-oxo,3-oxo-cyclobutanecarboxylicacid,1-carboxy-3-oxocyclobutane,3-oxocyclobutyl carboxylic acid,3-oxocyclobutanecarboxylicacid,pubchem14220,acmc-1cak7,ksc201q6h
4913358
3-oxocyclobutane-1-carboxylic acid
C1C(CC1=O)C(=O)O
23761-23-1
C5H6O3
114.1
MFCD00100900
IENOFRJPUPTEMI-UHFFFAOYSA-N
3-oxocyclobutanecarboxylic acid,3-oxo-cyclobutanecarboxylic acid,cyclobutanecarboxylic acid, 3-oxo,3-oxo-cyclobutanecarboxylicacid,1-carboxy-3-oxocyclobutane,3-oxocyclobutyl carboxylic acid,3-oxocyclobutanecarboxylicacid,pubchem14220,acmc-1cak7,ksc201q6h
4913358
3-oxocyclobutane-1-carboxylic acid
C1C(CC1=O)C(=O)O
Applications
Useful intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 5g
Color White
Melting Point 69°C to 70°C
Beilstein 2078401
Formula Weight 114.1
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 3-Oxocyclobutanecarboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning