Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Oxopentanenitrile, 99%

Manufacturer:  Alfa Aesar™ H50789MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50789MD


  Check Availability
Add to cart

Specifications

Description

Applications
3-Oxopentanenitrile is used as pharmaceutical intermediates.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.
Description & Specifications

Chemical Identifiers

33279-01-5
97.12
UN3439
3-Oxopentanenitrile
C5H7NO
MFCD07367087
Propionylacetonitrile

Specifications

33279-01-5
97.12
UN3439
27° to 30°C
3-Oxopentanenitrile
C5H7NO
MFCD07367087
Propionylacetonitrile
250mg
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit