Alfa Aesar™ 3-Oxopentanenitrile, 99%

Manufacturer: Alfa Aesar™ H5078903

Quantity 1g

 View more versions of this product

Catalog No. AAH5078903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 33279-01-5
    Molecular Formula C5H7NO
    Formula Weight 97.12
    MDL Number MFCD07367087
    UN Number UN3439
    Synonym Propionylacetonitrile
    Chemical Name or Material 3-Oxopentanenitrile

    Specifications

    Formula Weight 97.12
    Molecular Formula C5H7NO
    MDL Number MFCD07367087
    CAS 33279-01-5
    UN Number UN3439
    Synonym Propionylacetonitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H312-H332-H315-H319
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 27° to 30°C
    Quantity 1g
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 3-Oxopentanenitrile
    Assay 99%

    Description

    Applications
    3-Oxopentanenitrile is used as pharmaceutical intermediates.

    Notes
    Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.

    GHS H Statement
    H301-H312-H332-H315-H319
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit