Learn More
Thermo Scientific™ 3 Pack: Orion™ Star A211 pH Benchtop Meter Kit: PROMO
Measure pH, mV and temperature with the durable and dependable Thermo Scientific™ Orion Star™ A211 pH Benchtop Meter.
Specifications
AC Adapter Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Alarm Output High/low limit alarm, calibration due alarm
Battery Life 800 hr.
Battery Type 4 x AA (sold separately)
Calibration pH with calibration editing option, relative mV (RmV), ORP and temperature
View More Specs
Products 2
Catalog Number Mfr. No. Description Includes Price Quantity & Availability  
Catalog Number Mfr. No. Description Includes Price Quantity & Availability  
STARA2116P3
View Documents
Thermo Scientific™
STARA2116P3
Orion Star A211 pH benchtop meter kit with ROSS Ultra Triode epoxy-body pH/ATC electrode, solutions and stand Star A211 pH meter, 8157BNUMD ROSS Ultra Triode epoxy-body pH/ATC electrode, 810199 ROSS pH buffer and solution kit, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
STARA2117P3
View Documents
Thermo Scientific™
STARA2117P3
Orion Star A211 pH benchtop meter kit with general purpose pH/ATC electrode, solutions and stand Star A211 pH meter, 9157BNMD Triode epoxy-body pH/ATC electrode, 910199 pH buffer and solution kit, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Large LCD Display:
  • Plain language screen prompts, soft keys that update for easy selection
  • Multilanguage interface that supports English, Spanish, French, Italian, German and Chinese
  • AUTO-READ™ that locks in the stable reading on your screen, ready indicator alerts when readings are stable and timed reading gathers data in specific time intervals
Features:
  • Up to five point pH calibration with automatic recognition for USA/NIST™ and DIN buffers
  • Calibration editing for fixing calibration errors without a complete recalibration
  • Non-volatile memory to hold up to 2000 data points with time and date stamp
  • USB and RS232 ports and complimentary data analysis software to easily transfer data and keep meter software up-to-date
  • Direct control of the Orion Star stirrer probe (096019, sold separately) for mixing samples without a stir plate
Included:
  • Electrode arm and newly designed probe holder make it easier to maintain and place probes into samples
  • Universal power adapter works with almost every AC power source, or use four AA batteries (sold separately) to run the meter on DC power
  • IP54-rated housing handles splashes
  • Wall-mountable if table space is a problem
Specifications
Specifications
Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
800 hr.
pH with calibration editing option, relative mV (RmV), ORP and temperature
24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.)
7.000 pH
Non-volatile memory preserves data log, calibration log and meter settings
Universal AC adapter (included) or 4 AA batteries (sold separately)
High/low limit alarm, calibration due alarm
4 x AA (sold separately)
1
IP-54
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
10 per channel with password protection
3 Year Meter Warranty