Alfa Aesar™ 3-(Pentafluorothio)benzoic acid, 97%

Manufacturer: Alfa Aesar™ H3449906

Quantity 5g

 View more versions of this product

Catalog No. AAH3449906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 833-96-5
    Molecular Formula C7H5F5O2S
    Formula Weight 248.17
    MDL Number MFCD03788519
    UN Number UN2811
    Synonym 3-Carboxyphenylsulfur pentafluoride
    Chemical Name or Material 3-(Pentafluorothio)benzoic acid

    Specifications

    Formula Weight 248.17
    Melting Point 159° to 160°C
    Quantity 5g
    UN Number UN2811
    Synonym 3-Carboxyphenylsulfur pentafluoride
    Molecular Formula C7H5F5O2S
    MDL Number MFCD03788519
    CAS 833-96-5
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311
    Toxic if swallowed.
    Toxic in contact with skin.
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P361-P312-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 3-(Pentafluorothio)benzoic acid
    Assay 97%

    GHS H Statement
    H301-H311
    Toxic if swallowed.
    Toxic in contact with skin.

    GHS P Statement
    P280-P301+P310-P361-P312-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit