Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Penten-2-ol, 97%, Mixture of cis/trans (>90% trans), ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1569-50-2
C5H10O
86.134
MFCD00004544
GJYMQFMQRRNLCY-BHYDHMSTSA-N
e,2s-pent-3-en-2-ol,s-3-pentene-2-ol,s,e-3-pentene-2-ol,e,2s-3-penten-2-ol,s-trans-3-penten-2-ol,2s,3z-3-penten-2-ol,s-+-e-3-penten-2-ol,2s,3e-pent-3-en-2-ol
6999919
(E,2S)-pent-3-en-2-ol
CC=CC(C)O
1569-50-2
C5H10O
86.134
MFCD00004544
GJYMQFMQRRNLCY-BHYDHMSTSA-N
e,2s-pent-3-en-2-ol,s-3-pentene-2-ol,s,e-3-pentene-2-ol,e,2s-3-penten-2-ol,s-trans-3-penten-2-ol,2s,3z-3-penten-2-ol,s-+-e-3-penten-2-ol,2s,3e-pent-3-en-2-ol
6999919
(E,2S)-pent-3-en-2-ol
CC=CC(C)O
Density 0.8430g/mL
Color Colorless
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3CH=CHCH(OH)CH3
Beilstein 01, 443
Formula Weight 86.13
Physical Form Liquid
Percent Purity 96% (trans+cis, approximately), 90% (trans-isomer, approximately)
Chemical Name or Material 3-Penten-2-ol
Show More Show Less

GHS P Statement Flammable liquid and vapor.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a well-ventilated place. Keep container tightly closed.

Warning

EINECSNumber : 216-376-6

TSCA : TSCA