Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pentyn-1-ol, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
100g
Packaging:
Glass bottle
10229-10-4
C5H8O
84.12
MFCD00002956
IDYNOORNKYEHHO-UHFFFAOYSA-N
3-pentyn-1-ol,3-pentynol,acmc-1bxad
66295
pent-3-yn-1-ol
CC#CCCO
10229-10-4
C5H8O
84.12
MFCD00002956
IDYNOORNKYEHHO-UHFFFAOYSA-N
3-pentyn-1-ol,3-pentynol,acmc-1bxad
66295
pent-3-yn-1-ol
CC#CCCO
Specific Gravity 0.91
Refractive Index 1.4550 to 1.4570
Density 0.9100g/mL
Boiling Point 154.0°C to 157.0°C
Packaging Glass bottle
Quantity 5g
Flash Point 54°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3C≡CCH2CH2OH
Beilstein 01,IV,2227
Formula Weight 84.12
Percent Purity 99%
Chemical Name or Material 3-Pentyn-1-ol, 99%
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning

EINECSNumber : 233-550-7

TSCA : TSCA