Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3 Percent Platinum on Activated Carbon Powder, Reduced, Water Wet, 22 microns, BASF Catalyst, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
7440-06-4
Pt
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
[Pt]
7440-06-4
Pt
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
[Pt]
Additional Information Support: Activated Carbon powder
Particle Size 22 microns
Platinum (Pt) 0.03
Quantity 25 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.