Alfa Aesar™ 3-Phenoxy-1,2-propanediol, 98%

Manufacturer: Alfa Aesar™ B2570114

Quantity 25g

 View more versions of this product

Catalog No. AAB2570114

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 538-43-2
    Molecular Formula C9H12O3
    Formula Weight 168.19
    MDL Number MFCD00004717
    Synonym Phenyl glyceryl ether
    Chemical Name or Material 3-Phenoxy-1,2-propanediol

    Specifications

    Formula Weight 168.19
    Molecular Formula C9H12O3
    MDL Number MFCD00004717
    CAS 538-43-2
    Synonym Phenyl glyceryl ether
    RTECS Number TZ1050000
    Flash Point >110°C (230°F)
    Boiling Point 315°C
    Melting Point 56° to 60°C
    Quantity 25g
    Chemical Name or Material 3-Phenoxy-1,2-propanediol
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit