Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenoxy-1-propanol, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
6180-61-6
C9H12O2
152.19
MFCD00039549
AWVDYRFLCAZENH-UHFFFAOYSA-N
3-phenoxy-1-propanol, 1-propanol, 3-phenoxy, 3-phenoxy 1-propanol, unii-515pbp1u72, propoxylated phenol, ph 4p, polypropylene glycol phenyl ether, 3-phenoxypropanol, 3-phenyloxypropanol, polypropylene glycol monophenyl ether
80319
3-phenoxypropan-1-ol
C1=CC=C(C=C1)OCCCO
6180-61-6
C9H12O2
152.19
MFCD00039549
AWVDYRFLCAZENH-UHFFFAOYSA-N
3-phenoxy-1-propanol, 1-propanol, 3-phenoxy, 3-phenoxy 1-propanol, unii-515pbp1u72, propoxylated phenol, ph 4p, polypropylene glycol phenyl ether, 3-phenoxypropanol, 3-phenyloxypropanol, polypropylene glycol monophenyl ether
80319
3-phenoxypropan-1-ol
C1=CC=C(C=C1)OCCCO
Specific Gravity 1.04
Flash Point >112°C
Boiling Point 83°C to 85°C
Packaging Glass bottle
Refractive Index 1.5280 to 1.53
Density 1.0400g/mL
Color Colorless
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5O(CH2)3OH
Beilstein 06, 147
Formula Weight 152.19
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Phenoxy-1-propanol, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-228-8

TSCA : TSCA