Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenoxybenzaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
39515-51-0
C13H10O2
198.221
MFCD00003353
MRLGCTNJRREZHZ-UHFFFAOYSA-N
m-phenoxybenzaldehyde,3-phenoxy-benzaldehyde,benzaldehyde, 3-phenoxy,3-formyldiphenyl ether,m-phenyloxy benzaldehyde,3-phenoxy benzaldehyde,dsstox_cid_8005,dsstox_rid_78636,dsstox_gsid_28005,5-phenoxybenzaldehyde
38284
3-phenoxybenzaldehyde
C1=CC=C(C=C1)OC2=CC=CC(=C2)C=O
39515-51-0
C13H10O2
198.221
MFCD00003353
MRLGCTNJRREZHZ-UHFFFAOYSA-N
m-phenoxybenzaldehyde,3-phenoxy-benzaldehyde,benzaldehyde, 3-phenoxy,3-formyldiphenyl ether,m-phenyloxy benzaldehyde,3-phenoxy benzaldehyde,dsstox_cid_8005,dsstox_rid_78636,dsstox_gsid_28005,5-phenoxybenzaldehyde
38284
3-phenoxybenzaldehyde
C1=CC=C(C=C1)OC2=CC=CC(=C2)C=O
Refractive Index 1.5980 to 1.6000 (20°C, 589nm)
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Color Yellow to Amber
Density 1.1470g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5OC6H4CHO
Formula Weight 198.22
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 3-Phenoxybenzaldehyde, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 254-487-1

RTECSNumber : CU7560200

TSCA : TSCA