Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenoxypropionic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
7170-38-9
C9H10O3
166.176
MFCD00002769
BUSOTUQRURCMCM-UHFFFAOYSA-N
3-phenoxypropionic acid,propanoic acid, 3-phenoxy,beta-phenoxypropionic acid,3-penoxypropionic acid,3-phenoxy-propionic acid,3-phenoxypropionicacid,b-phenoxypropionate,3-phenoxypropionate,3-phenoxy-propanoate,beta-phenoxypropionate
81596
3-phenoxypropanoic acid
C1=CC=C(C=C1)OCCC(=O)O
7170-38-9
C9H10O3
166.176
MFCD00002769
BUSOTUQRURCMCM-UHFFFAOYSA-N
3-phenoxypropionic acid,propanoic acid, 3-phenoxy,beta-phenoxypropionic acid,3-penoxypropionic acid,3-phenoxy-propionic acid,3-phenoxypropionicacid,b-phenoxypropionate,3-phenoxypropionate,3-phenoxy-propanoate,beta-phenoxypropionate
81596
3-phenoxypropanoic acid
C1=CC=C(C=C1)OCCC(=O)O
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Color White to Beige
Melting Point 96°C to 100°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5OCH2CH2CO2H
Beilstein 06, 163
Formula Weight 166.18
Physical Form Fluffy Powder
Percent Purity ≥98.5%
Chemical Name or Material 3-Phenoxypropionic acid, 99%
Show More Show Less

EINECSNumber : 230-518-4

TSCA : TSCA