Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Phenoxythiophene, 97%

Click to view available options
Quantity:
1g
5g
25g
63285-84-7
C10H8OS
176.233
MFCD00178747
DXSHUWUDKNEGFS-UHFFFAOYSA-N
3-phenoxy-thiophene,acmc-20amz2
7018947
3-phenoxythiophene
C1=CC=C(C=C1)OC2=CSC=C2
63285-84-7
C10H8OS
176.233
MFCD00178747
DXSHUWUDKNEGFS-UHFFFAOYSA-N
3-phenoxy-thiophene,acmc-20amz2
7018947
3-phenoxythiophene
C1=CC=C(C=C1)OC2=CSC=C2
Quantity 25g
Formula Weight 176.23
Percent Purity 97%
Chemical Name or Material 3-Phenoxythiophene

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger