Alfa Aesar™ 3-Phenoxytoluene, 97%

Manufacturer: Alfa Aesar™ B2189922

Quantity 100g

 View more versions of this product

Catalog No. AAB2189922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3586-14-9
    Molecular Formula C13H12O
    Formula Weight 184.24
    MDL Number MFCD00008531
    Beilstein 2045714
    Synonym 3-Methyldiphenyl ether
    Chemical Name or Material 3-Phenoxytoluene

    Specifications

    Formula Weight 184.24
    TSCA TSCA
    Molecular Formula C13H12O
    MDL Number MFCD00008531
    Synonym 3-Methyldiphenyl ether
    RTECS Number DA6142000
    Flash Point >110°C (230°F)
    Boiling Point 271° to 273°C
    Quantity 100g
    CAS 3586-14-9
    Beilstein 2045714
    Chemical Name or Material 3-Phenoxytoluene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit