Alfa Aesar™ 3-Phenyl-1-propylboronic acid pinacol ester, 97%

Manufacturer: Alfa Aesar™ H28985MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH28985MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 329685-40-7
    Molecular Formula C15H23BO2
    Formula Weight 246.16
    MDL Number MFCD09953505
    Synonym 4,4,5,5-Tetramethyl-2-(3-phenylpropyl)-1,3,2-dioxaborolane
    Chemical Name or Material 3-Phenyl-1-propylboronic acid pinacol ester

    Specifications

    Formula Weight 246.16
    Molecular Formula C15H23BO2
    MDL Number MFCD09953505
    CAS 329685-40-7
    Synonym 4,4,5,5-Tetramethyl-2-(3-phenylpropyl)-1,3,2-dioxaborolane
    Quantity 250mg
    Chemical Name or Material 3-Phenyl-1-propylboronic acid pinacol ester
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit