Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenyl-3-pentanol, 97%

Manufacturer:  Alfa Aesar B2087106

Quantity 5g

 View more versions of this product

Catalog No. AAB2087106


  Check Availability
Add to cart

Specifications

Description

Applications
It finds its application in the synthesis and physical properties of some c12 and c13 phenylalkane and cyclohexylalkane hydrocarbons and in the dehydration of Alcohols in Dimethyl Sulfoxide1,2.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

1565-71-5
164.25
Diethyl phenyl carbinol
C11H16O
MFCD00021830
3-Phenyl-3-pentanol

Specifications

98° to 100°C (3mmHg)
1565-71-5
164.25
Diethyl phenyl carbinol
97%
5g
C11H16O
MFCD00021830
3-Phenyl-3-pentanol
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit