Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Phenylamino)cyclohex-2-ene-1-one, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
24706-50-1
C12H13NO
187.24
MFCD00082654
IVONJXTZXFTWNA-UHFFFAOYSA-N
3-anilino-2-cyclohexen-1-one,2-cyclohexen-1-one, 3-phenylamino,3-phenylamino cyclohex-2-en-1-one,3-phenylamino cyclohex-2-enone,3-anilinocyclohex-ene-1-one,3-phenylamino-2-cyclohexen-1-one,2-cyclohexen-1-one,3-phenylamino,acmc-1cr8y,3-anilino-2-cyclohexenone,aronis25001
98795
3-anilinocyclohex-2-en-1-one
C1CC(=CC(=O)C1)NC2=CC=CC=C2
24706-50-1
C12H13NO
187.24
MFCD00082654
IVONJXTZXFTWNA-UHFFFAOYSA-N
3-anilino-2-cyclohexen-1-one,2-cyclohexen-1-one, 3-phenylamino,3-phenylamino cyclohex-2-en-1-one,3-phenylamino cyclohex-2-enone,3-anilinocyclohex-ene-1-one,3-phenylamino-2-cyclohexen-1-one,2-cyclohexen-1-one,3-phenylamino,acmc-1cr8y,3-anilino-2-cyclohexenone,aronis25001
98795
3-anilinocyclohex-2-en-1-one
C1CC(=CC(=O)C1)NC2=CC=CC=C2
Packaging Glass Bottle
Melting Point 178°C to 182°C
Quantity 1g
Assay Percent Range 97.5% min. (HPLC)
Solubility Information Solubility in water: insoluble
Formula Weight 187.24
Percent Purity 98%
Chemical Name or Material 3-(Phenylamino)cyclohex-2-ene-1-one