Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Phenylazo-2,6-diaminopyridine hydrochloride, 98+%

Manufacturer:  Alfa Aesar™ B2188603

Quantity 1g

 View more versions of this product

Catalog No. AAB2188603


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

136-40-3
249.7
2,6-Diamino-3-(phenylazo)pyridine; Phenazopyridine hydrochloride
C11H11N5·HCl
MFCD00035347
3-Phenylazo-2,6-diaminopyridine hydrochloride

Specifications

1g
C11H11N5·HCl
MFCD00035347
≈240°C (dec.)
≥98%
136-40-3
249.7
2,6-Diamino-3-(phenylazo)pyridine; Phenazopyridine hydrochloride
3-Phenylazo-2,6-diaminopyridine hydrochloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H334-H351-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P301+P310-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : US7875000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit