Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Phenylphthalide, 99%

Click to view available options
Quantity:
5g
25g
5398-11-8
C14H10O2
210.23
MFCD00023099
SQFMIHCARVMICF-UHFFFAOYNA-N
3-phenylphthalide,3-phenyl phthalide,phthalide, 3-phenyl,3-phenyl-2-benzofuran-1 3h-one,3-phenyl-1 3h-isobenzofuranone,3-phenyl-1,3-dihydro-2-benzofuran-1-one,1 3h-isobenzofuranone, 3-phenyl,3-phenyl-3-hydroisobenzofuran-1-one,phenylphthalid,acmc-1ak9n
95365
3-phenyl-3H-2-benzofuran-1-one
C1=CC=C(C=C1)C2C3=CC=CC=C3C(=O)O2
5398-11-8
C14H10O2
210.23
MFCD00023099
SQFMIHCARVMICF-UHFFFAOYNA-N
3-phenylphthalide,3-phenyl phthalide,phthalide, 3-phenyl,3-phenyl-2-benzofuran-1 3h-one,3-phenyl-1 3h-isobenzofuranone,3-phenyl-1,3-dihydro-2-benzofuran-1-one,1 3h-isobenzofuranone, 3-phenyl,3-phenyl-3-hydroisobenzofuran-1-one,phenylphthalid,acmc-1ak9n
95365
3-phenyl-3H-2-benzofuran-1-one
C1=CC=C(C=C1)C2C3=CC=CC=C3C(=O)O2
Melting Point 116°C to 118°C
Quantity 25g
Beilstein 159678
Formula Weight 210.23
Percent Purity 99%
Chemical Name or Material 3-Phenylphthalide

EINECSNumber : 226-426-9

RTECSNumber : TI3800000

TSCA : No

Recommended Storage : Ambient temperatures