Alfa Aesar™ 3-Phenylpropionaldehyde, 95%

Manufacturer: Alfa Aesar™ A1036730

Quantity 250g

 View more versions of this product

Catalog No. AAA1036730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 104-53-0
    Molecular Formula C9H10O
    Linear Formula C6H5(CH2)2CHO
    Formula Weight 134.18
    MDL Number MFCD00007021
    Beilstein 1071910
    Synonym Hydrocinnamaldehyde; 3-Phenylpropanal
    Chemical Name or Material 3-Phenylpropionaldehyde

    Specifications

    Formula Weight 134.18
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 104-53-0
    Molecular Formula C9H10O
    Linear Formula C6H5(CH2)2CHO
    Synonym Hydrocinnamaldehyde; 3-Phenylpropanal
    Health Hazard 1 Warning
    Boiling Point 222° to 223°C
    Melting Point −42°C
    Flash Point 95°C (203°F)
    Quantity 250g
    TSCA TSCA
    MDL Number MFCD00007021
    Beilstein 1071910
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number MW4890000
    Chemical Name or Material 3-Phenylpropionaldehyde
    Assay 95%

    Description

    Applications
    3-Phenylpropionaldehyde is used as a flavoring agent. It is also used in the preparation of cinnamaldehyde, 2-chloro hydrocinnamaldehyde by alfa chlorination and mixture of homopropargyl alcohols. It serves as a precursor for the synthesis of mixture of syn- and anti-beta-hydroxyallylsilanes by hydroboration of allenylsilane. It is also involved in the catalytic asymmetric cyanosilylation to prepare (2S)-2-hydroxy-4-phenylbutanenitrile. Further, it undergoes Henry reaction to form nitroaldols by reacting with nitromethane.

    Solubility
    Miscible with chloroform, dichloromethane, ethyl acetate, alcohol and ether. Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents and strong bases.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit