Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Phenylpyridine, 97%

Click to view available options
Quantity:
5g
25g
1008-88-4
C11H9N
155.2
MFCD00006380
HJKGBRPNSJADMB-UHFFFAOYSA-N
pyridine, 3-phenyl,3-phenyl-pyridine,m-phenylpyridine,unii-edz6kb7jvc,5-phenylpyridine,3-phenyipyridine,edz6kb7jvc,3-azabiphenyl,3-pyridylbenzene,3-phenyl pyridine
13886
3-phenylpyridine
C1=CC=C(C=C1)C2=CN=CC=C2
1008-88-4
C11H9N
155.2
MFCD00006380
HJKGBRPNSJADMB-UHFFFAOYSA-N
pyridine, 3-phenyl,3-phenyl-pyridine,m-phenylpyridine,unii-edz6kb7jvc,5-phenylpyridine,3-phenyipyridine,edz6kb7jvc,3-azabiphenyl,3-pyridylbenzene,3-phenyl pyridine
13886
3-phenylpyridine
C1=CC=C(C=C1)C2=CN=CC=C2
Applications
3-Phenylpyridine is a useful synthetic intermediate. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Quantity 25g
Flash Point >110°C (230°F)
Boiling Point 272°C
Beilstein 110400
Formula Weight 155.2
Percent Purity 97%
Chemical Name or Material 3-Phenylpyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA