Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Picoline, 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
500mL
1L
2.5L
Packaging:
Glass bottle
108-99-6
C6H7N
93.13
MFCD00006402
ITQTTZVARXURQS-UHFFFAOYSA-N
3-picoline,beta-picoline,pyridine, 3-methyl,m-picoline,m-methylpyridine,b-picoline,beta-methylpyridine,.beta.-methylpyridine,.beta.-picoline,meta-methylpyridine
7970
CHEBI:39922
3-methylpyridine
CC1=CN=CC=C1
108-99-6
C6H7N
93.13
MFCD00006402
ITQTTZVARXURQS-UHFFFAOYSA-N
3-picoline,beta-picoline,pyridine, 3-methyl,m-picoline,m-methylpyridine,b-picoline,beta-methylpyridine,.beta.-methylpyridine,.beta.-picoline,meta-methylpyridine
7970
CHEBI:39922
3-methylpyridine
CC1=CN=CC=C1
Melting Point -19.0°C
Boiling Point 143.0°C to 144.0°C
Packaging Glass bottle
Specific Gravity 0.95
Density 0.9500g/mL
Flash Point 38°C
Refractive Index 1.5050 to 1.5070
Color Yellow
Quantity 1L
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 20, 239
Merck Index 15, 7513
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 93.13
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Picoline
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning