Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Picoline-N-oxide, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
1003-73-2
C6H7NO
109.13
MFCD00006203
DMGGLIWGZFZLIY-UHFFFAOYSA-N
3-methylpyridine 1-oxide,3-picoline-n-oxide,3-picoline n-oxide,3-methylpyridine n-oxide,3-methylpyridine-n-oxide,pyridine, 3-methyl-, 1-oxide,3-picoline 1-oxide,beta-picoline n-oxide,3-methylpyridin-1-ium-1-olate,3-methylpyridine-1-oxide
13858
3-methyl-1-oxidopyridin-1-ium
CC1=C[N+](=CC=C1)[O-]
1003-73-2
C6H7NO
109.13
MFCD00006203
DMGGLIWGZFZLIY-UHFFFAOYSA-N
3-methylpyridine 1-oxide,3-picoline-n-oxide,3-picoline n-oxide,3-methylpyridine n-oxide,3-methylpyridine-n-oxide,pyridine, 3-methyl-, 1-oxide,3-picoline 1-oxide,beta-picoline n-oxide,3-methylpyridin-1-ium-1-olate,3-methylpyridine-1-oxide
13858
3-methyl-1-oxidopyridin-1-ium
CC1=C[N+](=CC=C1)[O-]
Melting Point 38°C to 40°C
Flash Point 143°C
Boiling Point 150°C (15.0mmHg)
Color Brown to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: freely soluble in water
Formula Weight 109.13
Physical Form Crystalline Low Melting Mass
Percent Purity 98%
Chemical Name or Material 3-Picoline-N-oxide, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-714-4

RTECSNumber : UT5799000

TSCA : TSCA