Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Picolyl chloride hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
6959-48-4
C6H6ClN·HCl
164.04
MFCD00012818
UZGLOGCJCWBBIV-UHFFFAOYSA-N
3-chloromethyl pyridine hydrochloride,3-chloromethylpyridine hydrochloride,3-picolyl chloride hydrochloride,3-chloromethyl pyridine hcl,ccris 140,pyridine, 3-chloromethyl-, hydrochloride,3-pyridylmethyl chloride hydrochloride,unii-bqa8fyp4yn,3-chloromethyl pyridinium chloride,3-chloromethyl pyridine.hcl
23394
3-(chloromethyl)pyridine;hydrochloride
C1=CC(=CN=C1)CCl.Cl
6959-48-4
C6H6ClN·HCl
164.04
MFCD00012818
UZGLOGCJCWBBIV-UHFFFAOYSA-N
3-chloromethyl pyridine hydrochloride,3-chloromethylpyridine hydrochloride,3-picolyl chloride hydrochloride,3-chloromethyl pyridine hcl,ccris 140,pyridine, 3-chloromethyl-, hydrochloride,3-pyridylmethyl chloride hydrochloride,unii-bqa8fyp4yn,3-chloromethyl pyridinium chloride,3-chloromethyl pyridine.hcl
23394
3-(chloromethyl)pyridine;hydrochloride
C1=CC(=CN=C1)CCl.Cl
Melting Point 137°C to 143°C
Color White to Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: 820g/L
Formula Weight 164.04
Physical Form Crystalline Needles or Powder
Percent Purity ≥98.5%
Chemical Name or Material 3-Picolyl chloride hydrochloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
May cause damage to organs.
Harmful if swallowed.
Causes serious eye damage.
Suspected of causing cancer.
Suspected of causing genetic defects.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger