Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Piperidinecarboxylic acid, 97%

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Amber Glass Bottle
498-95-3
C6H11NO2
129.159
MFCD00005992
XJLSEXAGTJCILF-UHFFFAOYSA-N
nipecotic acid,3-piperidinecarboxylic acid,hexahydronicotinic acid,+/--nipecotic acid,h-dl-nip-oh,+/--3-piperidine carboxylic acid,dl-piperidine-3-carboxylic acid,nipecotic,h-nip-oh,3-carboxypiperidine
4498
CHEBI:116931
piperidine-3-carboxylic acid
C1CC(CNC1)C(=O)O
498-95-3
C6H11NO2
129.159
MFCD00005992
XJLSEXAGTJCILF-UHFFFAOYSA-N
nipecotic acid,3-piperidinecarboxylic acid,hexahydronicotinic acid,+/--nipecotic acid,h-dl-nip-oh,+/--3-piperidine carboxylic acid,dl-piperidine-3-carboxylic acid,nipecotic,h-nip-oh,3-carboxypiperidine
4498
CHEBI:116931
piperidine-3-carboxylic acid
C1CC(CNC1)C(=O)O
Infrared Spectrum Authentic
Quantity 50g
Color Beige-Yellow to White
Packaging Amber Glass Bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 129.16
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Piperidinecarboxylic acid, Functional Group amine (alkyl)
     carboxylic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.