Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotano 3-Pocket Reinforced Leather Large Primate Catch Glove™

Large Primate Catch Gloves isolate and protect fingers with a reinforced double layer of premium grade leather on the thumb and palm. A polyurethane insert above the wrist and forearm increases protection against bites, scratches, and lacerations.

$3825.98 - $8848.39

Specifications

Color Black
For Use With (Application) Protection Against Bites, Scratches, and Lacerations When Handling Large Primates
Glove Cuff Style Gauntlet
Grade Premium
Material Suede Leather
View More Specs

Products 9
Catalog Number Mfr. No. Hand Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Hand Quantity Price Quantity & Availability  
17-116-034
View Documents
Rotano
NB3POFALH20
Left 20 Gloves/Cs
Case for $3,825.98
Add to cart
 
17-116-031
View Documents
Rotano
NB3POFALH30
Left 30 Gloves/Cs
Case for $5,522.45
Add to cart
 
17-116-028
View Documents
Rotano
NB3POFALH50
Left 50 Gloves/Cs
Case for $8,848.39
Add to cart
 
17-116-033
View Documents
Rotano
NB3POFARH20
Right 20 Gloves/Cs
Case for $3,825.98
Add to cart
 
17-116-030
View Documents
Rotano
NB3POFARH30
Right 30 Gloves/Cs
Case for $5,522.45
Add to cart
 
17-116-027
View Documents
Rotano
NB3POFARH50
Right 50 Gloves/Cs
Case for $8,848.39
Add to cart
 
17-116-035
View Documents
Rotano
NB3POFAPR10
Specific 10 Pair/Cs
Case for $3,825.98
Add to cart
 
17-116-032
View Documents
Rotano
NB3POFAPR15
Specific 15 Pair/Cs
Case for $5,522.45
Add to cart
 
17-116-029
View Documents
Rotano
NB3POFAPR25
Specific 25 Pair/Cs
Case for $8,848.39
Add to cart
 
Description

Description

  • 3-Pocket style isolates and protects fingers while also maintaining dexterity
  • Second layer of full grain hide on back of hand and underneath wrist and forearm protects against bites, scratches, and lacerations
  • Polyurethane insert above the wrist and top of forearm for added coverage and impact resistance
  • Shoulder length upper arm/sleeve design prevents roll down while providing full arm protection and allowing for greater movement
  • Fleeced poly cotton liner for added insulation, sweat absorption, and comfort
  • High strength seam construction allows for added durability and strength
  • Non-exposed stitching to prevent snagging
  • Tested to EN 388:2016 + A1:2018 6.5 Puncture Resistance, Specifcation Force (N): Performance Level 4
Specifications

Specifications

Black
Gauntlet
Suede Leather
One Size
Keystone
Protection Against Bites, Scratches, and Lacerations When Handling Large Primates
Premium
Animal Handling Gloves
NB3P-OFA
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit